Contact Us
ImmunoCAP Rapid
Fast Answers for Fast Action
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Specific IgE Point-of-Care
ImmunoCAP™ Rapid™ is a quick and user-friendly point-of-care test to be performed in near-patient healthcare settings such as doctors’ offices and nurse stations to rule in or rule out allergy.1,2 It identifies 92 to 95 percent of symptomatic patients sensitized to any of the included allergens.3,4
There are three different test panels available, each holding a fixed set of 10 different allergens.*
ImmunoCAP Rapid
Wheeze/Rhinitis Child
Cat
Birch
Mugwort
Timothy
Egg White
Dog 
Olive pollen
Wall pellitory
House dust mite
Cow's milk
ImmunoCAP Rapid
Asthma/Rhinitis Adult
Cat
Birch
Mugwort
Timothy
Cockroach
Dog
Olive
Wall pellitory
House dust mite
Mold
ImmunoCAP Rapid
Allergen 8*
Cat
House dust mite
Cocksfoot
Common ragweed
Mugwort
Dog
Birch
Japanese Cedar
*ImmunoCAP Rapid Allergen 8 holds eight different allergens.
Single-use technology
ImmunoCAP Rapid is a single use, disposable testing device.
Broad application
The ImmunoCAP Rapid test can be performed irrespective of age, symptoms, disease activity, and antihistamine medication.
Easy and quick testing
ImmunoCAP Rapid is a specific IgE antibody blood test that will determine whether antibodies to any of the allergens included in the test are present in your patient’s blood. The test will provide a clear and reliable answer in 20 minutes while you wait.
Reliable results
You will receive proven and reliable results, with good concordance to ImmunoCAP Specific IgE blood tests.5
ImmunoCAP Rapid is available in dedicated European countries and in Japan.
Quickly rule in or rule out allergy
The test is a first-line evaluation tool that supports healthcare providers in the decision to rule in or rule out allergy.1,2
The test results should be interpreted in the context of the patient’s clinical history, before making a diagnosis. Regardless of the results, it is recommended that a symptomatic patient follow up with their healthcare provider.
ImmunoCAP Rapid Test Diagram
ImmunoCAP Rapid tests
View Catalog
Article Number 82-1000-01
Product Type Kit
Barcode BWF
Size 10 Devices
Product Info
Article Number 82-1001-01
Product Type Test
Barcode C46
Size 10 devices
Product Info
Article Number 82-1031-33
Product Type Test
Size 3 x 3 devices
Product Info
Explore the full ImmunoCAP portfolio.
View Portfolio
Find an instrument to run ImmunoCAP tests.
View Phadia Laboratory Systems
Get scientific information on whole allergens, allergen mixes, and allergen components.
Visit Allergen Encyclopedia
References
  1. Sarratud, T., et al., Accuracy of a point-of-care testing device in children with suspected respiratory allergy. Allergy Asthma Proc, 2010. 31(2): p. e11-7.
  2. Diaz-Vazquez, C., et al., Accuracy of ImmunoCAP Rapid in the diagnosis of allergic sensitization in children between 1 and 14 years with recurrent wheezing: the IReNE study. Pediatr Allergy Immunol, 2009. 20(6): p. 601-9.
  3. Hedlin, G., et al., Allergy Diagnosis in Children and Adults: Performance of a New Point-of-Care Device, ImmunoCAP Rapid. World Allergy Organ J, 2009. 2(7): p. 138-43.
  4. Eigenmann, P.A., et al., The ImmunoCAP Rapid Wheeze/Rhinitis Child test is useful in the initial allergy diagnosis of children with respiratory symptoms. Pediatr Allergy Immunol, 2009. 20(8): p. 772-9.
  5. Jan Forslid, P.M.L.S., Laboratory evaluation of a qualitative in vitro test for specific IgE in primary care as compared to quantitative measurement using automated ImmunoCAP FEIA processing. EAACI, 2008. Poster discussion 230.