These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
ImmunoCAP™ Rapid™ is a quick and user-friendly point-of-care test to be performed in near-patient healthcare settings such as doctors’ offices and nurse stations to rule in or rule out allergy.1,2 It identifies 92 to 95 percent of symptomatic patients sensitized to any of the included allergens.3,4
| ImmunoCAP Rapid Wheeze/Rhinitis Child |
Cat Birch Mugwort Timothy Egg White Dog Olive pollen Wall pellitory House dust mite Cow's milk |
| ImmunoCAP Rapid Asthma/Rhinitis Adult |
Cat Birch Mugwort Timothy Cockroach Dog Olive Wall pellitory House dust mite Mold |
| ImmunoCAP Rapid Rhinitis/Asthma 1* |
Cat Mold Cocksfoot Common ragweed Mugwort Dog Cockroach Japanese Cedar
|
*ImmunoCAP Rapid Rhinitis/Asthma 1 holds eight different allergens.
ImmunoCAP Rapid is a single use, disposable testing device.
The ImmunoCAP Rapid test can be performed irrespective of age, symptoms, disease activity, and antihistamine medication.
ImmunoCAP Rapid is a specific IgE antibody blood test that will determine whether antibodies to any of the allergens included in the test are present in your patient’s blood. The test will provide a clear and reliable answer in 20 minutes while you wait.
You will receive proven and reliable results, with good concordance to ImmunoCAP Specific IgE blood tests.5
ImmunoCAP Rapid is available in dedicated European countries and in Japan.
The test is a first-line evaluation tool that supports healthcare providers in the decision to rule in or rule out allergy.1,2
The test results should be interpreted in the context of the patient’s clinical history, before making a diagnosis. Regardless of the results, it is recommended that a symptomatic patient follow up with their healthcare provider.