Technical Resources - Media Formulations
11120 - MEM Vitamin Solution (100X) liquid
ComponentsMolecular WeightConcentration (mg/L)mM
Vitamins
Choline chloride139.62100.00.7162298
D-Calcium pantothenate477.0100.00.2096436
Folic Acid441.0100.00.22675736
Nicotinamide122.13100.00.8187997
Pyridoxal hydrochloride203.62100.00.4911109
Riboflavin376.3710.00.0265696
Thiamine hydrochloride337.27100.00.29649836
i-Inositol180.16200.01.1101243
Inorganic Salts
Sodium Chloride (NaCl)58.448500.0145.44833
参照
Related Products:
11130 - MEM Amino Acids Solution (50X) liquid
Cat No: 11130036 , 11130051 , 11130077
11140 - MEM Non-Essential Amino Acids Solution
Cat No: 11140050 , 11140076