Technical Resources - Media Formulations
11140 - MEM Non-Essential Amino Acids Solution
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.0750.010.0
L-Alanine89.0890.010.0
L-Asparagine132.01320.010.0
L-Aspartic acid133.01330.010.0
L-Glutamic Acid147.01470.010.0
L-Proline115.01150.010.0
L-Serine105.01050.010.0
参照
Related Products:
11120 - MEM Vitamin Solution (100X) liquid
Cat No: 11120052
11130 - MEM Amino Acids Solution (50X) liquid
Cat No: 11130036 , 11130051 , 11130077