先天性免疫異常症(IEI)
IEIが何を示すのか、原発性免疫不全症候群(PID)との関連や表現型の分類について
先天性免疫異常症(IEI)とは
近年、「IEI(Inborn Errors of Immunity)」という用語を目にすることが増えています。ある文献では、「自己免疫疾患や自己炎症性疾患、アレルギーや悪性腫瘍などに関連する幅広い表現型を捉えるために、【先天性免疫異常症】(IEI)という用語が提案された」1と報告されています。
国際免疫学会連合(International Union of Immunological Societies, IUIS)の専門委員会はIEIの遺伝子型と表現型分類を発表しました2,3。IUIS専門委員会は病因をもとに400以上の疾患を整理し、10の表に分類しました。これらの表にはそれぞれ報告済みの遺伝子変異も記載されていますが、その数は年々増加しています。
  •  Table I - Immunodeficiencies affecting cellular and humoral immunity
  •  Table II - Combined immunodeficiencies with associated or syndromic features
  •  Table III - Predominantly antibody deficiencies
  •  Table IV - Diseases of immune dysregulation
  •  Table V - Congenital defects of phagocyte (number or function)
  •  Table VI - Defects in intrinsic and innate immunity
  •  Table VII - Autoinflammatory diseases
  •  Table VIII - Complement deficiencies
  •  Table IX - Bone marrow failure
  •  Table X - Phenocopies of inborn errors of immunity
文献では、先天性免疫異常症(IEI)と原発性免疫不全症候群(PID)という用語はしばしば同じ意味で使われていますが、現状では、IEIよりもPIDという用語の方が好まれる場合もあるようです4。PIDからIEIに名称を変更する、あるいはこれらの用語を互換的に使用するという議論もありますが、いくつか注意が必要な点もあります。例えば、PID患者の大部分を占める疾患のうち、3種類の疾患(IgA欠損症、分類不能型免疫不全症(CVID)、乳児一過性低ガンマグロブリン血症)は現在のところ遺伝的要因が定義されていません。また、無ガンマグロブリン血症や重症複合免疫不全症(SCID)のような明確に定義された表現型であっても、すべての患者に遺伝的説明がつくわけではありません4
2015年から2020年にかけて、先天性免疫異常症の原因として同定された遺伝子異常の数は、ほぼ2倍に増えています2
原発性免疫不全症候群(PID)
関連疾患
原発性免疫不全症候群(PID)
原発性免疫不全症候群(PID)の徴候や症状、疾患例、診断について
イムノアッセイ
製品情報
イムノアッセイ
単クローン性ガンマグロブリン血症や免疫システムの異常を呈する患者さんの診断補助にお役立ていただける各種アッセイをご紹介します
References
1. Notarangelo LD, et al. Human inborn errors of immunity: An expanding universe. Science immunology 2020; 5:eabb1662
2. Tangye SG, et al. Human Inborn Errors of Immunity: 2019 Update on the Classification from the International Union of Immunological Societies Expert Committee. J Clin Immunol 2020; 40:24-64
3. Tangye SG, Al-Herz W, Bousfiha A, Chatila T, Cunningham-Rundles C, Etzioni A, Franco JL, Holland SM, Klein C, Morio T, Ochs HD, Oksenhendler E, Picard C, Puck J, Torgerson TR, Casanova JL, Sullivan KE. Human Inborn Errors of Immunity: 2019 Update on the Classification from the International Union of Immunological Societies Expert Committee. J Clin Immunol. 2020 Jan;40(1):24-64. doi: 10.1007/s10875-019-00737-x. Epub 2020 Jan 17. Erratum in: J Clin Immunol. 2020 Feb 22;: PMID: 31953710; PMCID: PMC7082301.
4. Ameratunga R, Longhurst H, Lehnert K, Steele R, Edwards ESJ, Woon ST. Are All Primary Immunodeficiency Disorders Inborn Errors of Immunity? Front Immunol. 2021 Jul 21;12:706796. doi: 10.3389/fimmu.2021.706796. PMID: 34367167; PMCID: PMC8335567.
お問い合わせについて
当社の製品・サービスに関するご質問は、 お問い合わせフォームからご連絡ください
Photo of lab staff member pipetting