Multiple Myeloma Diagnosis Criteria for Primary Care Providers
What are the symptoms to look out for?
Introduction
Why is a timely diagnosis important?
More than 50% of Myeloma patients
Experience a delay in diagnosis of more than 6 months when diagnosed at primary care1.
More than 70% of patients
Experience additional symptoms and complications as a result of a delayed diagnosis2.
Timely diagnosis and subsequent management of Multiple Myeloma avoids complications that impact patients’ quality of life. Earlier diagnosis and therefore commencement of treatment significantly improves 5-year survival rates of patients.
Primary care providers therefore play a key role in recognizing and ruling out Multiple Myeloma at the earlier stages of the disease.
5 Year Survival Rate
84%
Earlier Stage Diagnosis
26%
Advanced Stage Diagnosis
Speaker
Featured in this video
Professor Andrew Spencer
Head of Malignant Haematology, Stem Cell Transplantation and Head of Myeloma Research at The Alfred Hospital, Australia
Key Takeaways
Recognize early warning signs of Multiple Myeloma
Multiple Myeloma symptoms are non-specific and may mimic the aging process or more common conditions encountered at primary care, such as diabetes, hypertension and cardiac disease4.
Patients often present at primary care with bone pain, back pain, aches and generally feeling unwell.
When symptoms are unexplained or persist despite intervention, look out for one or more early warning signs of Multiple Myeloma4.
What are the early warning signs and blood tests to order?
Recognize
early warning signs
Request
correct blood tests
Refer
to hematologist
CRAB events of Multiple Myeloma
As Multiple Myeloma progresses, advanced presentations are associated with end-organ damage. These are referred to as CRAB events of Multiple Myeloma and include hypercalcemia, renal impairment, anaemia, and bone lesions.
Request the correct blood tests to rule out Multiple Myeloma
A number of blood tests are recommended to rule out Multiple Myeloma, as highlighted in the publication by Dr Joseph Mikhael, MD and Chief Medical Officer, International Myeloma Foundation4. This includes—although not limited to—the following:
  • Complete Blood Count: to check for anemia and raised erythrocyte sedimentation rate (ESR)
  • Serum Biochemistry Panel: to check for raised calcium, raised creatinine and low albumin
  • Serum Protein Studies: to check for M-protein using serum protein electrophoresis and serum free light chain (sFLC) tests
Adapted from:
Seesaghur A et al. Clinical features and diagnosis of multiple myeloma: a population-based cohort study in primary care. BMJ Open 20213
Refer to Hematology
When the patient symptoms and basic laboratory findings are suggestive of Multiple Myeloma, assessment of serum protein studies will help to rule out Multiple Myeloma4.
The survival rate for Multiple Myeloma patients is increased by over one and a half times when the diagnosis is achieved through the primary care referral pathway- rather that the emergency route5.
As the gatekeepers to earlier diagnosis of Multiple Myeloma, primary care practitioners should be alert to the early warning signs. Requesting the best test combination to rule out Multiple Myeloma earlier, allows for a timelier referral pathway to hematology.
What are the early warning signs and blood tests to order?
If you liked this content, you will also like these:
Understanding Multiple Myeloma
Article
Understanding Multiple Myeloma
Symptoms, manifestations and complications
Multiple Myeloma Monitoring Guidelines for Relapse Detection
Expert Insight
Multiple Myeloma Monitoring Guidelines for Relapse Detection
The International Myeloma Working Group (IMWG) produce guidelines for monitoring Multiple Myeloma and related diseases
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you