Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment?  Visit our
Troubleshooting page
General
Relative quantitation compares the amount of an analyte in different samples. Absolute quantitation measures the actual amount of an analyte in a sample using a known standard at different concentrations. Label-free, SILAC, and TMT (Tandem Mass Tag) workflows are used for relative quantitation of protein samples. Heavy (i.e., AQUA) peptides are used as standard for absolute quantitation of target proteins.
Tandem Mass Tag technology is used to individually label different protein samples so they can be combined into a single sample for LC-MS analysis. The major advantages of this workflow are higher sample throughput, less instrument analysis time, high precision of peptide quantitation among replicates, and fewer missing quantified proteins among different samples.
We recommend using high-resolution Orbitrap Tribrid (e.g. Orbitrap Ascend or Orbitrap Eclipse), Orbitrap Exploris (e.g., 240, 480), or Q Exactive Plus series of instruments.
SILAC can be used to multiplex 2 to 3 samples. TMT reagents can be used to multiplex 2, 6, 10, and 11 samples. TMTpro reagents can be used to measure up to 16-18 samples simultaneously. 32 samples can be measured using a combination of TMTpro non-deuterated and deuterated tags.
Label-free, TMT reagents, and TMTpro reagents can be used with any protein sample type. SILAC and Neucode quantitation can only be used with cell lines and model organisms that can be metabolically labeled.
TMT reagents and TMTpro reagents are different chemicals with different masses. Therefore, we do not recommend mixing the two tags as the combined samples would be significantly more complex resulting in fewer quantified proteins.
The LC-MS are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only. Not for use in diagnostic procedures.
CMD SchemaApp code