On this page:
Consistent, accurate assay performance—right out of the box
The Applied Biosystems resDNASEQ Residual Host Cell DNA kit offers a complete sample-to-results solution, including a highly-characterized genomic DNA standard*, for accurate validation of sensitivity and specificity. The kits are designed to detect fragmented and unfragmented DNA resulting in confident quantitation. Developed using rich bioinformatics and a sophisticated primer/probe design pipeline, then internally validated to help meet performance standards, resDNASEQ assays deliver highly accurate and consistent results kit-to-kit, lot-to-lot, and year-to-year.
resDNASEQ kits include:
resDNASEQ kits were designed to meet or exceed regulatory guidelines.
Why choose a commercial kit for residual DNA testing?
The overall performance of the resDNASEQ method, including sensitivity, accuracy, precision, range, and linearity meets or exceeds the example described in USP <509>.
Implementation of a fully integrated testing solution can reduce the expense and effort in procurement of assay components from multiple vendors and qualification of critical reagents that is required for in-house developed assays.
resDNASEQ quantitative DNA portfolio
The resDNASEQ residual host cell DNA quantitation system offers a robust portfolio of real-time and digital PCR assay kits to measure residual DNA levels from a large variety of host cell lines—including the plasmid DNA kanamycin resistance gene—used in the production of biopharmaceutical products. 
resDNASEQ Quantitative 
CHO DNA kit
qPCR    dPCR
resDNASEQ Quantitative
E. coli DNA kit
 qPCR    dPCR
resDNASEQ Quantitative
HEK293 DNA kit
 qPCR
resDNASEQ Quantitative
Human DNA kit
 qPCR
resDNASEQ Quantitative
MDCK DNA kit
 qPCR
resDNASEQ Quantitative
NS0 DNA kit
 qPCR
resDNASEQ Quantitative
Pichia pastoris DNA kit
qPCR
resDNASEQ Quantitative
Sf9/Baculovirus DNA kit
 qPCR    dPCR
resDNASEQ Quantitative
Vero DNA kit
 qPCR
resDNASEQ Quantitative
E1A DNA Fragment Length kit
qPCR    dPCR
The Applied Biosystems resDNASEQ Quantitative E1A DNA Fragment Length Kit can be used in combination with the resDNASEQ Quantitative HEK293 DNA Kit to measure the length of residual E1A DNA fragments that are transformed into the HEK293 cell line. The workflow can use the same sample as for the resDNASEQ Quantitative HEK293 DNA Kit.
Precision in practice – guidance for analytical excellence
In this quick 60-second video (give or take), our field application scientist tackles your top FAQ about residual host cell DNA testing using the resDNASEQ kit. It speaks to the essentials: ensuring your standard curve quality, mastering DNA extraction from complex samples, running triplicate recovery controls, and using an internal PCR control to catch any issues. Get the answers you need, fast!
Discover our residual plasmid DNA quantitation solution
On-demand webinar: Quantitating Residual Host Cell and Plasmid DNA in a Biotherapeutic Manufacturing Workflow
Fast Facts: Robust quantitation of residual host cell and plasmid DNA & oncogenic fragments in HEK-based viral vector manufacturing
On-demand webinar: Simplify residual DNA quantitation in viral vector production: qPCR and dPCR tools
Request a technical consultation
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Shop DNA quantitation kits
Catalog # Name Size Price Qty
4402085 Each
3.678.528
4413713 Each
4.293.282
A59366 Each -
4458435 Each
3.678.528
4460366 Each
4.293.282
A57017 Each -
A46014 Each
3.578.568
A46565 Each
4.098.360
A26366 Each -
A27335 Each -
4464335 Each
3.678.528
4464339 Each -
4458441 Each
3.778.488
4460364 Each -
4464336 Each
3.778.488
4464340 Each
4.098.360
A46066 Each
5.097.960
A47405 Each
5.717.712
A59319 Each -
A41797 Each
3.573.570
A45522 Each
4.098.360
A51969 Each -
A51970 Each -
A55852 Each -
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
For Research Use Only. Not for use in diagnostic procedures.