resDNASEQ™ dPCR E1A DNA Fragment Length Kit
Applied Biosystems™

resDNASEQ™ dPCR E1A DNA Fragment Length Kit

The resDNASEQ dPCR E1A DNA Fragment Length Kit is an absolute quantitation PCR -based system that measures the fragment lengthsRead more
Have Questions?
Catalog NumberQuantity
A55852100 reactions
Catalog number A55852
Price (USD)
3,920.00
Each
Quantity:
100 reactions
Price (USD)
3,920.00
Each
The resDNASEQ dPCR E1A DNA Fragment Length Kit is an absolute quantitation PCR -based system that measures the fragment lengths of residual host-cell DNA by targeting the E1A gene in HEK 293 cell lines, which are predominantly used in the development of gene therapies, cell-based vaccines, and similar biotherapeutics. Reliable and rapid, the resDNASEQ kit enables highly sensitive and specific quantitation of different fragment lengths of the E1A gene within the residual host-cell DNA. This helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types—from in-process samples with different sample matrices to purified final product—whether the sample contains high molecular weight or sheared DNA.

Features of the resDNASEQ dPCR E1A DNA Fragment Length Kit include:
• Consistent performance across the expected range of DNA fragment sizes
• Simple— streamlined workflow for ease-of-use with your digital PCR instrument
• Fast—minimal hands-on time; results in ∼90 minutes when used with QuantStudio Absolute Q Digital PCR System
• Reliable—consistent performance across expected range of DNA fragment sizes

Integrated workflow solution
The resDNASEQ dPCR E1A DNA Fragment Length Kit is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing. The resDNASEQ kit has been validated on the QuantStudio Absolute Q Digital PCR System.

The Absolute Q system has a high tolerance of common inhibitors in biopharmaceutical sample matrices, eliminating the need for nucleic acid extraction in most cases. If nucleic acid extraction is required, we recommend use of the PrepSEQ Residual DNA Sample Preparation Kit, which offers both a manual and automated sample preparation workflow.

Easy to use
With the resDNASEQ dPCR E1A DNA Fragment Length Kit, you perform dPCR analysis to quantitate the copy numbers of three different fragment size ranges in your test samples. The resDNASEQ kit includes almost everything needed to carry out these steps, from master mix to TaqMan primer/probe mix to E1A DNA standards, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of fragments of three different lengths of E1A DNA, assay results must be unaffected by the size of non-specific DNA present in the test sample. With the resDNASEQ system you get consistent assay performance across the three E1A fragment sizes. 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Host CellHEK and other Cell Lines transformed by E1A
No. of Reactions100 reactions
Product LineresDNASEQ
Product TypeDigital PCR Kit
Quantity100 reactions
Shelf Life12 months
Shipping ConditionDry Ice
PCR MethodDigital PCR
Unit SizeEach
Contents & Storage
resDNASEQ E1A DNA Control
• E1A DNA Control, 0.12 mL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ E1A DNA dPCR Reagents
• Absolute Q Digital PCR Master Mix, 5X, 2 x 360 μL, store at –15 to –25°C, protect from light
• E1A DNA Assay Mixes (short, medium, long), 220 μL each, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use