Search
Studying the brain and neuro-functions requires the knowledge of a vast selection of experimental methods, from cell preparation to image acquisition and analysis. Thermo Scientific Amira Software supports you in the most frequently used image analysis techniques, such as filament tracing and editing, DTI analysis, brain perfusion analysis, and object tracking. Combining Amira Software's versatility with state-of-the-art 3D visualization and image processing enables you to create custom workflows that extract exactly the desired type of information from an image.
Explore industry-leading insights and research that can support your lab workflow. Amira Software can empower your lab with a cutting-edge, comprehensive imaging data analysis toolbox.
Helping scientists answer questions that enable breakthrough discoveries in life sciences, materials science, and industry.
![]() | Correlative Microscopy: Using Amira Software to Understand the Spread of Cancer Read more |
![]() | Amira Software Accelerates Veterinary Research with 3D Visualization and Analysis Read more |
![]() | Molecular Clarity—Discovering What’s Possible with Cryo-Electron Tomography Read more |
![]() | Cryo-Electron Tomography and 3D Software Advances Coronavirus Research Read more |
Understanding the interconnectivity of the brain is essential for functional brain research. This allows researchers to better understand how different parts of the brain jointly orchestrate higher cognitive functions and motor skills.
With Amira Software, you are able to perform the entire DTI analysis workflow by first converting images into Talairach coordinates. You are then able to map multiple brain images onto a reference brain. Once the brain images are aligned, gradient images can be converted into a tensor field, and nerve fiber bundles can be tracked and visualized with our state-of-the-art visualization tools.
With Amira Software, you are able to trace, analyze, and quantify 3D images of filamentous structures such as neurons and blood vessels. Filamentous networks can be reconstructed, and measures such as length, thickness, orientation, ranks, etc., can be computed and visualized in compelling 3D renderings. Tracing can be performed automatically or interactively, depending on the application and needs.
Our template-matching algorithm allows automatic detection and tracing of fine filaments in noisy cryo-EM data. Amira Software also enables you to edit the resulting graphs to remove image features erroneously identified as a filament or to add missing parts of a network.
Brain perfusion studies help researchers and scientist to understand the impact of strokes or other brain tissue diseases such as Alzheimer and dementia. The exact location and duration of a stroke can determine functional impact of the event.
With Amira Software, you are able to analyze brain perfusion in perfusion-weighted MRI and CT images. This analysis includes computation of mean transit time (MTT), cerebral blood flow (CBF), and cerebral blood volume (CBV).
In vitro neuroscience research has grown considerably in neurodegenerative, neurogenesis, and neurotoxicology applications. Neurite outgrowth and synaptogenesis are the gold standards for evaluating health and functionality of neurons. However, even in vitro, identification of neuronal outgrowth morphology is difficult.
Researchers manually tracing neurites and synaptogenic spots face variability and non-scalability. Amira Software’s three-dimensional neuroscience abilities can accurately and efficiently quantify neuronal morphology in 3D models to facilitate the high-throughput demands of this research.
Understanding the interconnectivity of the brain is essential for functional brain research. This allows researchers to better understand how different parts of the brain jointly orchestrate higher cognitive functions and motor skills.
With Amira Software, you are able to perform the entire DTI analysis workflow by first converting images into Talairach coordinates. You are then able to map multiple brain images onto a reference brain. Once the brain images are aligned, gradient images can be converted into a tensor field, and nerve fiber bundles can be tracked and visualized with our state-of-the-art visualization tools.
With Amira Software, you are able to trace, analyze, and quantify 3D images of filamentous structures such as neurons and blood vessels. Filamentous networks can be reconstructed, and measures such as length, thickness, orientation, ranks, etc., can be computed and visualized in compelling 3D renderings. Tracing can be performed automatically or interactively, depending on the application and needs.
Our template-matching algorithm allows automatic detection and tracing of fine filaments in noisy cryo-EM data. Amira Software also enables you to edit the resulting graphs to remove image features erroneously identified as a filament or to add missing parts of a network.
Brain perfusion studies help researchers and scientist to understand the impact of strokes or other brain tissue diseases such as Alzheimer and dementia. The exact location and duration of a stroke can determine functional impact of the event.
With Amira Software, you are able to analyze brain perfusion in perfusion-weighted MRI and CT images. This analysis includes computation of mean transit time (MTT), cerebral blood flow (CBF), and cerebral blood volume (CBV).
In vitro neuroscience research has grown considerably in neurodegenerative, neurogenesis, and neurotoxicology applications. Neurite outgrowth and synaptogenesis are the gold standards for evaluating health and functionality of neurons. However, even in vitro, identification of neuronal outgrowth morphology is difficult.
Researchers manually tracing neurites and synaptogenic spots face variability and non-scalability. Amira Software’s three-dimensional neuroscience abilities can accurately and efficiently quantify neuronal morphology in 3D models to facilitate the high-throughput demands of this research.
Amira、Avizo、およびPerGeosソフトウェアの新規ユーザー用に設計された初級者向けトレーニングは学習速度を高め、投資を最大化します。
このコースは講義と実践セッションで構成されています。トレーニング教材は、Amira、Avizo、およびPerGeosソフトウェアの基本的な特長や機能を取り上げています。
Amira、Avizo、およびPerGeosソフトウェアの既存ユーザー用に設計された上級者向けトレーニングは投資を最大化し、結果までの時間の短縮します。
このコースは講義と実践セッションで構成されています。トレーニング教材は、Amira、Avizo、およびPerGeosソフトウェアの高度な特長や機能を取り上げています。
サーモフィッシャーサイエンティフィックには25年以上にわたる3Dおよび画像処理の経験と、大小の規模の組織に提供してきたカスタムプロジェクトの実績があり、お客様の独自のニーズに合わせてソリューションを提供できます。
当社のソフトウェアソリューションをさまざまなレベルでカスタマイズおよび拡張できます。
Visualization of the results of a Diffusion Tensor Imaging study of the human head Courtesy of Prof. Dr. Alexander Brawanski, University Hospital of Regensburg1001.
Visualization of a brain of Mus musculus.
Fertilizers effects on neuronal outgrowth and synapse expression by IUF – Leibniz Research Institute for Environmental Medicine.
Visualization of the results of a Diffusion Tensor Imaging study of the human head Courtesy of Prof. Dr. Alexander Brawanski, University Hospital of Regensburg1001.
Visualization of a brain of Mus musculus.
Fertilizers effects on neuronal outgrowth and synapse expression by IUF – Leibniz Research Institute for Environmental Medicine.
- バイオフォーマット
- ビットマップフォーマット
- 顕微鏡:電子および光学
- 医学および神経画像フォーマット
- 分子フォーマット
- その他の画像取得機器(MRI、X線など)
For Research Use Only. Not for use in diagnostic procedures.