Potassium • Alkali Metal
Primary XPS region: K2p
Overlapping regions: C1s
Binding energies of common chemical states:
Chemical stateBinding energy K2p3/2
KCl292.9 eV
KBr293.2 eV
Charge referenced to adventitious C1s peak at 284.8eV.
Experimental information
  • It is often quicker to acquire C1s region and extend range to 305eV to capture the K2p region.
Interpretation of XPS spectra
  • K2p peak has clearly spaced spin-orbit components (Δchloride=2.8eV).
Strong And Weak Potassium Peaks Versus Carbon
Join-the-conversation-banner-in-blogs-social-graphic-1143250-1000x172
Potassium • Alkali Metal
  • Weaker K2p peaks can be easily confused with either C1s (CF2) peaks or C1s satellite structure in aromatic compounds.
    • Assignment of K2p versus C1s feature is possible by identifying spin-orbit components expected for K2p.
    • Also check for K2s peak (~378eV).
General comments
  • Since potassium is a highly reactive metal, it is always present as a compound, e.g., oxide.
Element Crystal bcc
About this element
Symbol: K
Date of discovery: 1807
Name origin: English potash
Appearance: silvery
Discoverer: Sir Humphry Davy
Obtained from: carnallite
Melting point: 336.53 K
Boiling point: 1032 K
Density[kg/m3]: 856
Molar volume: 45.94 × 10-6 m3/mol
Protons/Electrons: 19
Neutrons: 20
Shell structure: 2,8,8,1
Electron configuration: [Ar]4s1
Oxidation state: 1
Crystal structure: cubic body centered
Potassium, the second lightest metal, was discovered by Sir Humphry Davy in 1807. It reacts violently with water and can even catch fire spontaneously in water. It can be found bound to other elements in seawater and minerals, and is needed for plant growth. Potassium is commonly used in fertilizer. It is also used in gunpowder and glass, and plays a vital role in animal cells as one of the partners in the Na-K pump. K-40 decays into Ar-40 and this process is used in the dating of rocks.
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.