Technical Resources - Media Formulations
LB Agar, powder (Lennox L Agar)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Sodium Chloride (NaCl)58.05000.086.206894
Other Components
SELECT Agar, bacteriological12000.0Infinity
SELECT Peptone 140 (pancreatic digest of casein)10000.0Infinity
SELECT Yeast Extract autolyzed low sodium5000.0Infinity
参照
Related Products:
LB Broth Base, powder (Lennox L Broth Base)
Cat No: 12780029 , 12780052
LB Broth (1X) liquid
Cat No: 10855021