Technical Resources - Media Formulations
20012 - PBS, pH 7.2
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Phosphate monobasic (KH2PO4)136.0210.01.5441177
Sodium Chloride (NaCl)58.09000.0155.17241
Sodium Phosphate dibasic (Na2HPO4-7H2O)268.0726.02.7089553
参照
Related Products:
70013 - PBS, 10x, pH 7.2
Cat No: 70013016 , 70013032 , 70013065 , 70013073
10010 - PBS, pH 7.4
Cat No: 10010001 , 10010002 , 10010015 , 10010023 , 10010031 , 10010049 , 10010056 , 10010072
70011 - PBS,10x, pH 7.4
Cat No: 70011036 , 70011044 , 70011051 , 70011069