Technical Resources - Media Formulations
B-27 Supplement Minus Insulin (50X) liquid
Contains no insulin.
B-27 Serum-Free Supplement is used for growth and long-term viability of hippocampal neurons. This supplement is to be used with a combination of basal medium such as Neurobasal Medium. Below is the composition list. The concentrations are proprietary.
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Vitamins
Biotinconfidentialn/a
DL Alpha Tocopherol Acetateconfidentialn/a
DL Alpha-Tocopherolconfidentialn/a
Vitamin A (acetate)confidentialn/a
Proteins
BSA, fatty acid free Fraction Vconfidentialn/a
Catalaseconfidentialn/a
Human Transferrinconfidentialn/a
Superoxide Dismutaseconfidentialn/a
Other Components
Corticosteroneconfidentialn/a
D-Galactoseconfidentialn/a
Ethanolamine HClconfidentialn/a
Glutathione (reduced)confidentialn/a
L-Carnitine HClconfidentialn/a
Linoleic Acidconfidentialn/a
Linolenic Acidconfidentialn/a
Progesteroneconfidentialn/a
Putrescine 2HClconfidentialn/a
Sodium Seleniteconfidentialn/a
T3 (triodo-I-thyronine)confidentialn/a
参照
Related Products:
B-27 Serum-Free Supplement (50X) liquid
Cat No: 17504001 , 17504044
B-27 Supplement Minus AO (50X) liquid
Cat No: 10889038
B-27 Supplement Minus Vitamin A (50X) liquid
Cat No: 12587001 , 12587010