Protocols
Related Product Information
Catalog # Name Size Price (USD) Qty
AM1333 MEGAscript™ T7 Transcription Kit, 25 Reactions Each
303.65
Online Exclusive
312.00
Save 8.35 (3%)
Protocol
Below is a protocol for making biotinylated amplified RNA (aRNA) using one of Ambion's MEGAscript™ Kits. The reaction incorporates 2 different biotinylated nucleotides for maximum sensitivity.
Each 20 µl reaction should contain:
1.5µlds cDNA template
2µl75 mM ATP
2µl75 mM GTP
1.5µl75 mM CTP
1.5µl75 mM UTP
3.75µl10 mM Bio-11-CTP
3.75µl10 mM Bio-16-UTP or Bio-11-UTP
2µl10X reaction buffer
2µlT7 RNA Polymerase
20µltotal volume
Assemble the reaction in the order shown. Do not assemble the reaction on ice, since spermidine in the reaction buffer can lead to precipitation of the template DNA. If multiple reactions are needed a "master mix" can be prepared by multiplying the volume of each reagent by the number of reactions.
Mix the reaction by flicking the tube and briefly microfuge to collect all of the reaction mixture at the bottom of the tube. Incubate at 37°C for 6 hrs or more (overnight incubation may increase yields). After incubation, add 1 µl DNase I (included in the MEGAscript Kit) to remove the DNA template. Incubate at 37°C for 30 min. Purify the cRNA by precipitation or by using an Ambion RNAqueous™ column. Biotinylated rNTPs can be obtained from the following suppliers:
PerkinElmer NEN
NEL542 Bio-11-CTP 10 mM, 250 nmol in 25 µl
NEL543  Bio-11-UTP 10 mM, 250 nmol in 25 µl
ENZO
42815 Bio-11-UTP 10 mM, 250 nmol in 25 µl
42818 Bio-11-CTP 10 mM, 250 nmol in 25 µl
TOP
LT036