Single-Use Fermentor System
On this page:
Designed specifically for microbial fermentation
The Thermo Scientific HyPerforma Enhanced Single-Use Fermentor (e.S.U.F.) is helping to pioneer the single-use technology market by offering a rigorous solution for microbial fermentation applications while featuring the flexibility, ease of use, and efficiency found in single-use systems.
The e.S.U.F. is engineered for optimal oxygen mass transfer, mixing, and temperature control, with performance comparable to conventional stainless-steel systems. S.U.F.s use fundamental engineering principles, helping to make scale-up and process development work readily streamlined. Available in 30 L and 300 L sizes, the e.S.U.F. consists of a fermentor tank and a standard Thermo Scientific HyPerforma e.S.U.F. BioProcess Container (BPC).
For those who value speed and performance
The HyPerforma e.S.U.F. facilitates fast turnaround times and reliable fermentation. It offers purpose-built performance in line with stainless steel fermenters—allowing you to enjoy the benefits of a single-use system, including lower capital investment, quicker turnaround, lower contamination potential, and lower facility, energy, and water requirements than most other systems on the market.
Better mixing, oxygen uptake, and cooling performance
The HyPerforma e.S.U.F. features enhanced parabolic turbine impellers that deliver oxygen uptake comparable to or better than stainless steel fermentors. Additional features include:
  • Purpose-built fermentor design that can outperform modified bioreactors offered by our competition
  • Enhanced jacketing includes 35% more cooling surface area, offering sufficient cooling to handle high-density fermentations
  • An extensive library of application data includes both internal and customer-generated performance data
Quality and experience
  • Patented BPC design for optimal performance
  • High-quality consumables with a very low failure rate
  • With 6–300 L working volumes, the e.S.U.F. offers a flexible closed system designed to eliminate cross-contamination
Features of eS.U.F. and standard S.U.F.
Developed with standard stainless-steel fermentation design geometry (3:1 height to diameter, 5:1 turndown ratio), the e.S.U.F. and standard S.U.F. support scale-up and process development activities.
Feature highlights
  • 3:1 total height-to-width aspect ratio for optimal cooling surface-to-volume ratio
  • The BPCs are highly configurable, allowing for flexibility to address specific application needs
  • Complete mixing system with a water jacket for temperature control
  • Drive shaft inserts into the BPC through the mixing drive motor and locks into the BPC agitator assembly of the S.U.F. BPC
  • Integrally sealed ports in the S.U.F. BPC allow for the addition of sensor probes and line sets
  • Integrated foam sensor
  • Single-use pH, DO, temperature, foam, and pressure sensors with a clear optical density plug port
  • Exhaust management system with options of low or high flow hydrophobic filter membranes and multiple vent filters based on gas flow needs 
  • High flow exhaust condenser with back-up filters, gas sparge filter line sets
  • Feed, submerge fill, syringe sample, and drain line set options
Powerful agitation facilitates high oxygen transfer (kLa) and a homogeneous reaction
  • 3 Rushton impellers with standard S.U.F. or enhanced parabolic with eS.U.F.
  • Impellers couple directly to the shaft through a bearing to drive the motor
  • 4 baffles
  • Drilled-hole sparge (DHS) with ring-like design
  • Innovative exhaust management and condenser systems are designed to provide reliable off-gassing and help minimize liquid loss during the fermentation process
System options
  • Exhaust condenser unit (extend filter life)
  • Exhaust gas vent filter heater (extend filter life, recommended 151 W for 300 L)
  • Backup filter, automated pinch clamp (extend filter life)
  • Load cells 
  • Cable/tubing management tree
  • Turnkey system with HyPerforma G3 process control system
  • Electrical box for remote agitation or open-architecture control
All S.U.F.s require a separate external temperature control unit and controller. You may choose an open-architecture approach or a turnkey, ready-to-use S.U.F. system.
Benefits of single-use fermentors
Enhanced HyPerforma Single-Use Fermentor Systems (eS.U.F.)
Enhancements to improve the performance of single-use fermentors were made to the 300 L hardware, as well as the 30 L and 300 L BPCs, to more closely match the oxygen delivery and cooling capacity performance of stainless steel fermentors.
Hardware enhancements
The cooling jacket surface area was increased on the sides and bottom for the 300 L S.U.F. to support adequate cooling of the production process. This 35% increase in jacketed surface area enables faster cooling and supports high-density cultures.
This image compares the hardware and jacketing surface of the HyPerforma S.U.F. (300 L) and HyPerforma eS.U.F. (300 L).
High-quality consumables designed for superior performance and reliability
Fermentor BPCs are made with robust Thermo Scientific Aegis5-14 film, offering exceptional reliability and backed by decades of experience.
The Thermo Scientific BioTitan Retention Device is an innovative replacement for traditional cable ties that enhances the reliability and integrity of assemblies with a 360-degree seal.
Next-generation centrifuge-ready BPCs provide a single-use, sterilized, closed system for cell harvesting in bioprocessing.
BioProcessing Container (BPC) enhancements
The performance of the eS.U.F. system hinges on the BPC and its associated impellers. The BPC incorporates three advanced parabolic turbine impellers, specifically engineered to optimize mass transfer in fermentation processes. These impellers have demonstrated a fourfold increase in oxygen delivery compared to standard Ruston impellers. Additionally, the BPC features our trusted Aegis film and the BioTitan retention device, which, combined with decades of single-use experience, help ensure a reliable and robust BPC.
Automation software and controllers
The HyPerforma S.U.F. automation system integrates with the Thermo Scientific TruBio software, powered by the DeltaV™ Live Platform and the Thermo Scientific HyPerforma G3 bioprocess controllers to offer easy, unified data aggregation and quality control, helping to improve the user experience.
Interested in leasing or financing our equipment?
Manage your resources by taking advantage of financing and leasing options from Thermo Fisher Scientific.
Single-Use Bioprocessing Equipment and Automation
We are committed to delivering exceptional performance, reliability, and quality.
Single-Use Bioprocessing
Our innovative single-use technologies enable you to accelerate biological manufacturing processes to create flexibility and operate more dynamically within upstream and downstream workflows.
BioProcessing Containers and Nalgene Bottles
We offer high-quality storage, transport, and sampling solutions, including Thermo Scientific BioProcess Containers (BPCs) and Thermo Scientific Nalgene containers.
Speak to a specialist
Connect with a Bioprocessing Single-Use Technology representative.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.