Technical Resources - Media Formulations
22100 - Glasgow's MEM, powder
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
L-Arginine hydrochloride211.042.00.19905214
L-Cystine 2HCl313.031.00.09904154
L-Glutamine146.0292.02.0
L-Histidine hydrochloride-H2O210.021.00.1
L-Isoleucine131.052.00.39694658
L-Leucine131.052.00.39694658
L-Lysine hydrochloride183.073.00.3989071
L-Methionine149.015.00.10067114
L-Phenylalanine165.033.00.2
L-Threonine119.047.60.39999998
L-Tryptophan204.08.00.039215688
L-Tyrosine disodium salt dihydrate261.052.00.19923371
L-Valine117.046.80.4
Vitamins
Choline chloride140.02.00.014285714
D-Calcium pantothenate477.02.00.004192872
Folic Acid441.02.00.0045351475
Niacinamide122.02.00.016393442
Pyridoxal hydrochloride204.02.00.009803922
Riboflavin376.00.25.319149E-4
Thiamine hydrochloride337.02.00.005934718
i-Inositol180.03.60.02
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0200.01.8018018
Ferric Nitrate (Fe(NO3)3"9H2O)404.00.12.4752476E-4
Magnesium Sulfate (MgSO4-7H2O)120.097.670.8139166
Potassium Chloride (KCl)75.0400.05.3333335
Sodium Chloride (NaCl)58.06400.0110.344826
Sodium Phosphate monobasic (NaH2PO4-H2O)138.0124.00.89855075
Other Components
D-Glucose (Dextrose)180.04500.025.0
Phenol Red376.415.00.039851222
参照
1. House, W., (1964) Medical Research Council, Institute of Virology,  University of Glasgow, Scotland, (November). 
2. MacPherson, I. and Stoker, M., (1962)  Virology 16:147.
Related Products:
11710 - Glasgow's MEM
Cat No: 11710035