Search
View additional product information for eBioscience™ ELISA/ELISPOT Diluent (5X) - FAQs (00-4202-56)
1 product FAQs found
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.