Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis)
Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis)
Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis)
Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis)
Thermo Scientific Chemicals

Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
数量:
50 x 100 mm
50 x 500 mm
製品番号(カタログ番号) 000097.FY
または、製品番号000097-FY
価格(JPY)
-
見積もりを依頼する
数量:
50 x 100 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
さらに表示
FormFoil, 0.25 mm (0.01 in.) thick
Total Metal Impurities≤0.001%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only