Gold powder, spherical, <10 micron, 99.95% (metals basis)
Gold powder, spherical, &lt;10 micron, 99.95% (metals basis)
Gold powder, spherical, &lt;10 micron, 99.95% (metals basis)
Gold powder, spherical, &lt;10 micron, 99.95% (metals basis)
Thermo Scientific Chemicals

Gold powder, spherical, <10 micron, 99.95% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
製品番号(カタログ番号) 000765.06
または、製品番号000765-06
価格(JPY)
-
お問い合わせください ›
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
さらに表示
FormPowder
Total Metal Impurities≤0.05%
Particle SizeD95: <10 microns
Surface Area0.1-0.35 m²/g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only