UltraComp eBeads™ Plus Compensation Beads
Invitrogen™

UltraComp eBeads™ Plus Compensation Beads

UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry. Fluorochrome-conjugated antibody binds자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호테스트 수
01-3333-42100
01-3333-4125
카탈로그 번호 01-3333-42
제품 가격(KRW)
492,000
온라인 행사
Ends: 30-Sep-2026
578,000
할인액 86,000 (15%)
Each
테스트 수:
100
제품 가격(KRW)
492,000
온라인 행사
Ends: 30-Sep-2026
578,000
할인액 86,000 (15%)
Each

UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry. Fluorochrome-conjugated antibody binds the spherical particles for use as a single-color compensation controls. Like the first generation beads, they bind with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies. They are designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers. Unlike the first generation beads, UltraComp eBeads Plus compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.

Features include:
• Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
• Improved fluorophore compatibility

Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody. When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed. This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.

RUO use only.
사양
농도5 x 106 beads/mL
직경(미터법)5 μm
여기/방출UV-633/635 nm
용도(애플리케이션)Mouse, rat, hamster, human, rabbit antibodies
테스트 수100
수량100 tests
제품 유형Compensation Beads
Unit SizeEach

자주 묻는 질문(FAQ)

What is the size of UltraComp eBeads Plus Compensation Beads?

The bead size is 5 µm.