Thermo Scientific Chemicals

Beryllium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010001.14
25 g, Each
Thermo Scientific Chemicals

Beryllium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010001.14
25 g, Each
Quantity

Chemical Identifiers

CAS
1304-56-9
IUPAC Name
oxoberyllium
Molecular Formula
BeO
InChI Key
LTPBRCUWZOMYOC-UHFFFAOYSA-N
SMILES
[Be]=O
Total Metal Impurities
1% max.

Description

Beryllium oxide is widely used in industry due to its low density and high thermal and electrical conductivity. It is used in high-technology ceramics, electronic heat sinks, electrical insulators, high-power devices, high-density electrical circuits, microwaves, X-ray windows, nuclear reactor fuels and for various aerospace and military applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text