Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)
Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)
Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)
Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)
Thermo Scientific Chemicals

Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)

CAS: 7440-36-0 | Sb | 121.76 g/mol
数量:
100 g
500 g
2 kg
製品番号(カタログ番号) 010095.A3
または、製品番号010095-A3
価格(JPY)
-
見積もりを依頼する
数量:
2 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-36-0
IUPAC Nameantimony
Molecular FormulaSb
InChI KeyWATWJIUSRGPENY-UHFFFAOYSA-N
SMILES[Sb]
さらに表示
Appearance (Color)Silver to gray
Total Metal Impurities0.5% max.
FormLump
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only