Thermo Scientific Chemicals

Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals

Catalog number: 010130.30
250 g, Each
Thermo Scientific Chemicals

Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals

Catalog number: 010130.30
250 g, Each
Quantity

Chemical Identifiers

CAS
7782-42-5
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Trace Metal
75-82% C, 18-25% Ash
Particle Size
-325 mesh (less than 45 micron)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text