Rubidium, 99.75% (metals basis) - FAQs
Rubidium, 99.75% (metals basis) - FAQs
View additional product information for Rubidium, 99.75% (metals basis) - FAQs (010315.03, 010315.14, 010315.06, 010315.22)
1 product FAQs found
What type of container is Rubidium 99.75% (metal basis) sold in?
Cat. No. 044214.xx Rubidium 99.75% (metal basis) is ampouled under Argon.
Cat. No. 010315.xx Rubidium 99.75% (metal basis) is vacuum sealed in a breakseal ampoule.