Platinum(II) 2,4-pentanedionate, Pt 48.0% min
Platinum(II) 2,4-pentanedionate, Pt 48.0% min
Platinum(II) 2,4-pentanedionate, Pt 48.0% min
Thermo Scientific Chemicals

Platinum(II) 2,4-pentanedionate, Pt 48.0% min

CAS: 15170-57-7 | C10H14O4Pt | 393.30 g/mol
製品番号(カタログ番号) 010526.06
または、製品番号010526-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15170-57-7
IUPAC Nameplatinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O4Pt
InChI KeyKLFRPGNCEJNEKU-FDGPNNRMSA-L
SMILES[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
さらに表示
Assay (unspecified)Pt 48.0% min
Used for photoactivated hydrosilylation reactions between vinyl and silane groups; in the deposition of platinum metal and in the preparation of platinum nanotubes. Precursor for FePt superlattice magnetic nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only