Thermo Scientific Chemicals

Potassium tetracyanoplatinate(II) trihydrate, 99.9% (metals basis), Pt 44.9% min, Thermo Scientific Chemicals

Catalog number: 010536.06
5 g, Each
Thermo Scientific Chemicals

Potassium tetracyanoplatinate(II) trihydrate, 99.9% (metals basis), Pt 44.9% min, Thermo Scientific Chemicals

Catalog number: 010536.06
5 g, Each
Quantity
Catalog number: 010536.06
Price (USD)
Price: 787.00
Online price: 668.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14323-36-5
IUPAC Name
dipotassium tetracyanoplatinumbis(ylium) trihydrate
Molecular Formula
C4H6K2N4O3Pt
InChI Key
KBALGZKVNFJSRU-UHFFFAOYSA-N
SMILES
O.O.O.[K+].[K+].N#C[Pt++](C#N)(C#N)C#N
Appearance (Color)
White to pale yellow
Form
Crystalline
Assay (special)
Pt 44.9-45.9%
Total Metal Impurities
≤0.1%

Description

Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text