Thermo Scientific Chemicals

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010661.18
50 g, Each
Thermo Scientific Chemicals

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010661.18
50 g, Each
Quantity
Catalog number: 010661.18
also known as 010661-18
Price (USD)

Chemical Identifiers

CAS
654054-66-7
IUPAC Name
cadmium(2+) dichloride
Molecular Formula
CdCl2
InChI Key
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cd++]
Total Metal Impurities
0.002% max.

Description

Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text