Thermo Scientific Chemicals

Cadmium hydroxide, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010662.09
10 g, Each
Thermo Scientific Chemicals

Cadmium hydroxide, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010662.09
10 g, Each
Quantity
Catalog number: 010662.09
also known as 010662-09
Price (USD)

Chemical Identifiers

CAS
21041-95-2
IUPAC Name
cadmium(2+) dihydroxide
Molecular Formula
CdH2O2
InChI Key
PLLZRTNVEXYBNA-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Cd++]
Total Metal Impurities
0.02% max.

Description

It is found in the industrial nickel cadmium storage batteries, in cadmium plating, in the manufacture of cadmium salts, laboratory reagents. It is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text