Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号)数量
010721.14
または、製品番号010721-14
25 g
製品番号(カタログ番号) 010721.14
または、製品番号010721-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS598-63-0
IUPAC Nameλ²-lead(2+) carbonate
Molecular FormulaCO3Pb
InChI KeyMFEVGQHCNVXMER-UHFFFAOYSA-L
SMILES[Pb++].[O-]C([O-])=O
さらに表示
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only