Lithium metaborate, Puratronic™, 99.997% (metals basis)
Lithium metaborate, Puratronic™, 99.997% (metals basis)
Lithium metaborate, Puratronic™, 99.997% (metals basis)
Thermo Scientific Chemicals

Lithium metaborate, Puratronic™, 99.997% (metals basis)

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 010739.36
または、製品番号010739-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
さらに表示
Total Metal Impurities0.003% max.
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only