Nickel(II) nitrate hexahydrate, Puratronic™, 99.9985% (metals basis)
Nickel(II) nitrate hexahydrate, Puratronic™, 99.9985% (metals basis)
Nickel(II) nitrate hexahydrate, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Nickel(II) nitrate hexahydrate, Puratronic™, 99.9985% (metals basis)

CAS: 13478-00-7 | H12N2NiO12 | 290.79 g/mol
数量:
5 g
50 g
250 g
製品番号(カタログ番号) 010816.18
または、製品番号010816-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13478-00-7
IUPAC Namenickel(2+) hexahydrate dinitrate
Molecular FormulaH12N2NiO12
InChI KeyAOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Assay (special)±2.5 % of Theoretical value (Ni)
FormCrystalline powder or granules
Appearance (Color)Green to blue-green or deep-green
Total Metal Impurities≤0.0015% (15 ppm)
It is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only