Thermo Scientific Chemicals

Selenium(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010854.14
25 g, Each
Thermo Scientific Chemicals

Selenium(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010854.14
25 g, Each
Quantity
Catalog number: 010854.14
also known as 010854-14
Price (USD)
Price: 294.00
Online price: 249.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7446-08-4
IUPAC Name
(oxo-λ⁴-selanylidene)oxidane
Molecular Formula
O2Se
InChI Key
JPJALAQPGMAKDF-UHFFFAOYSA-N
SMILES
O=[Se]=O
Total Metal Impurities
0.001% max.

Description

In organic reactions, selenium dioxide is often used as an oxidizing reagent employed for carrying out allylic oxidations, Riley oxidation, synthesis of diones, etc. It is used to impart a red color to glass. It finds use as a toner in photographic developing, and an active ingredient in some cold-blueing solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text