Thermo Scientific Chemicals

Potassium nitrate, 99.994% (metals basis), Thermo Scientific Chemicals

Catalog number: 011008.14
25 g, Each
Thermo Scientific Chemicals

Potassium nitrate, 99.994% (metals basis), Thermo Scientific Chemicals

Catalog number: 011008.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 011008.14
also known as 011008-14
Price (USD)
Price: 82.70
Your price:
Quantity
-

Chemical Identifiers

CAS
7757-79-1
IUPAC Name
potassium nitrate
Molecular Formula
KNO3
InChI Key
FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES
[K+].[O-][N+]([O-])=O
Total Metal Impurities
0.006% max.

Description

Potassium nitrate has several applications. It is used as an oxidizer, food preservative, fertilizer, and an active ingredient of condensed aerosol fire suppression systems. It is a powerful nitrating agent in sulfuric acid medium. In addition, it also used as an electrolyte in a salt bridge and thermal storage medium in power generation systems. Ternary salts containing potassium nitrate have been reported to improve the heat storage capacity in the molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text