Sodium chloride, Optical Grade
Sodium chloride, Optical Grade
Sodium chloride, Optical Grade
Thermo Scientific Chemicals

Sodium chloride, Optical Grade

CAS: 7647-14-5 | ClNa | 58.44 g/mol
製品番号(カタログ番号) 011019.18
または、製品番号011019-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
仕様
沸点1413°C
CAS7647-14-5
ChEBICHEBI:26710
化学物質名または材質Sodium chloride
White
さらに表示
Sodium Chloride (NaCl) is widely used in biochemistry and molecular biology applications. It is considered an essential nutrient. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. NaCl is a component of phosphate buffered saline and SSC buffer. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only