Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min
Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min
Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min
Thermo Scientific Chemicals

Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min

CAS: 13820-41-2 | Cl4H8N2Pt | 372.96 g/mol
製品番号(カタログ番号) 011046.03
または、製品番号011046-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13820-41-2
IUPAC Nameplatinum(2+) diammonium tetrachloride
Molecular FormulaCl4H8N2Pt
InChI KeyQJIMNDWDOXTTBR-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]
さらに表示
Total Metal Impurities0.1% max., Pt 51% min
Ammonium tetrachloroplatinate(II) is used in spectral analysis standard. It is also used in the preparation of platinum sponge and platinum catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only