Thermo Scientific Chemicals

Thulium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011201.09
10 g, Each
Thermo Scientific Chemicals

Thulium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011201.09
10 g, Each
Quantity
Catalog number: 011201.09
also known as 011201-09
Price (USD)
Price: 627.00
Online price: 532.65
Your price:
Quantity
-

Chemical Identifiers

CAS
19423-86-0
IUPAC Name
thulium(3+) trichloride
Molecular Formula
Cl3Tm
InChI Key
ILOTUXNTERMOJL-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Tm+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Thulium(III) chloride hydrate is used extremely as a rare metal to study the metal science of thulium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text