n-Tetracontane, 97% min
n-Tetracontane, 97% min
n-Tetracontane, 97% min
Thermo Scientific Chemicals

n-Tetracontane, 97% min

CAS: 4181-95-7 | C40H82 | 563.10 g/mol
製品番号(カタログ番号) 011274.04
または、製品番号011274-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4181-95-7
IUPAC Nametetracontane
Molecular FormulaC40H82
InChI KeyKUPLEGDPSCCPJI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
さらに表示
Appearance (Color)White
Assay (GC)≥97.0%
FormSolid
n-Tetracontane is used in a passivation layer in copper-phthalocyanine field-effect transistors for improving the charge carrier mobility and used to modify the quasi two-dimensional surface electron system on Au(111).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only