Europium(III) oxide, REacton™, 99.99% (REO)
Europium(III) oxide, REacton™, 99.99% (REO)
Europium(III) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Europium(III) oxide, REacton™, 99.99% (REO)

CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol
数量:
1 g
5 g
25 g
100 g
製品番号(カタログ番号) 011299.03
または、製品番号011299-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1308-96-9
IUPAC Namedieuropium(3+) trioxidandiide
Molecular FormulaEu2O3
InChI KeyAEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Eu+3].[Eu+3]
さらに表示
Total Rare Earth Oxide Impurities0.01% max.
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only