Thermo Scientific Chemicals

Cerium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011325.30
250 g, Each
Thermo Scientific Chemicals

Cerium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011325.30
250 g, Each
Quantity
Catalog number: 011325.30
also known as 011325-30
Price (USD)
Price: 168.00
Online price: 142.65
Your price:
Quantity
-

Chemical Identifiers

CAS
19423-76-8
IUPAC Name
cerium(3+) trichloride
Molecular Formula
CeCl3
InChI Key
VYLVYHXQOHJDJL-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ce+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text