Thermo Scientific Chemicals

Tantalum(V) oxide, 99.85% (metals basis), Thermo Scientific Chemicals

Catalog number: 011336.14
25 g, Each
Thermo Scientific Chemicals

Tantalum(V) oxide, 99.85% (metals basis), Thermo Scientific Chemicals

Catalog number: 011336.14
25 g, Each
Quantity
Catalog number: 011336.14
also known as 011336-14
Price (USD)
Price: 133.00
Online price: 113.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-61-0
IUPAC Name
[(dioxotantalio)oxy]dioxotantalum
Molecular Formula
O5Ta2
InChI Key
PBCFLUZVCVVTBY-UHFFFAOYSA-N
SMILES
O=[Ta](=O)O[Ta](=O)=O
Appearance (Color)
White to pale cream
Form
Powder
Total Metal Impurities
≤0.15% (1500 ppm)
Particle Size
-325 Mesh

Description

Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text