Indium shot, 4-8mm (0.2-0.3in), Puratronic™, 99.9999% (metals basis)
Indium shot, 4-8mm (0.2-0.3in), Puratronic™, 99.9999% (metals basis)
Indium shot, 4-8mm (0.2-0.3in), Puratronic™, 99.9999% (metals basis)
Thermo Scientific Chemicals

Indium shot, 4-8mm (0.2-0.3in), Puratronic™, 99.9999% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
数量:
100 Pc.
25 Pc.
製品番号(カタログ番号) 011359.KN
または、製品番号011359-KN
価格(JPY)
-
数量:
100 Pc.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
さらに表示
Total Metal Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only