Thermo Scientific Chemicals

Magnesium titanium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011398.A1
1 kg, Each
Thermo Scientific Chemicals

Magnesium titanium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011398.A1
1 kg, Each
Quantity
Catalog number: 011398.A1
also known as 011398-A1
Price (USD)
Price: 113.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12032-30-3
IUPAC Name
titanium(4+) magnesium(2+) trioxidandiide
Molecular Formula
MgO3Ti
InChI Key
XSETZKVZGUWPFM-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Mg++].[Ti+4]
Total Metal Impurities
1% max.

Description

Magnesium titanium oxide is used in the primary chemical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text