Thermo Scientific Chemicals

Strontium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011399.A1
1 kg, Each
Thermo Scientific Chemicals

Strontium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011399.A1
1 kg, Each
Quantity
Catalog number: 011399.A1
also known as 011399-A1
Price (USD)
Price: 145.00
Online price: 123.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12060-59-2
Total Metal Impurities
0.9% max.

Description

Strontium titanium oxide is used as a diamond stimulant and also used as a strontium-90 in radioisotope thermoelectric generators. It is used in the electronics and electrical insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text