Thermo Scientific Chemicals

Potassium perrhenate, 99% (metals basis), Re 64%, Thermo Scientific Chemicals

Catalog number: 011411.09
10 g, Each
Thermo Scientific Chemicals

Potassium perrhenate, 99% (metals basis), Re 64%, Thermo Scientific Chemicals

Catalog number: 011411.09
10 g, Each
Quantity
Catalog number: 011411.09
also known as 011411-09
Price (USD)
Price: 277.00
Online price: 235.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10466-65-6
IUPAC Name
potassium rheniumoylolate
Molecular Formula
KO4Re
InChI Key
QFKRWIFGDGKWLM-UHFFFAOYSA-N
SMILES
[K+].[O-][Re](=O)(=O)=O
Total Metal Impurities
1% max., Re 64%

Description

A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text