Thermo Scientific Chemicals

Silver bromide, 99.5%, Thermo Scientific Chemicals

Catalog number: 011425.06
5 g, Each
Thermo Scientific Chemicals

Silver bromide, 99.5%, Thermo Scientific Chemicals

Catalog number: 011425.06
5 g, Each
Quantity
Catalog number: 011425.06
also known as 011425-06
Price (USD)
Price: 40.30
Online price: 34.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7785-23-1
IUPAC Name
silver(1+) bromide
Molecular Formula
AgBr
InChI Key
ADZWSOLPGZMUMY-UHFFFAOYSA-M
SMILES
[Br-].[Ag+]
Appearance (Color)
Yellow/green
Assay (special)
99.5-100.0%
Form
Powder

Description

Silver bromide is used as the light-sensitive component on ordinary photographic film and plates and also used in I.R. applications and ionic semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text