Nickel rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis)
Nickel rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis)
Nickel rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis)
Thermo Scientific Chemicals

Nickel rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
数量:
10 cm
50 cm
150 cm
製品番号(カタログ番号) 011453.BS
または、製品番号011453-BS
価格(JPY)
-
数量:
10 cm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
さらに表示
Assay from Supplier's CofA≥99.5% (metals basis)
Appearance (Color)Silvery
FormRod, 12.7mm (0.5in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only