Vanadium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
Vanadium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
Vanadium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Vanadium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
製品番号(カタログ番号) 011483.G5
または、製品番号011483-G5
価格(JPY)
-
数量:
5 m
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
さらに表示
FormWire, 0.127mm (0.005in) diameter.
Appearance (Color)White silver
Assay from Supplier's CofA≥99.8% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only