Antimony(III) fluoride, 99+%
Antimony(III) fluoride, 99+%
Antimony(III) fluoride, 99+%
Antimony(III) fluoride, 99+%
Thermo Scientific Chemicals

Antimony(III) fluoride, 99+%

CAS: 7783-56-4 | F3Sb | 178.755 g/mol
製品番号(カタログ番号) 011492.36
または、製品番号011492-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7783-56-4
IUPAC Nametrifluorostibane
Molecular FormulaF3Sb
InChI KeyGUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILESF[Sb](F)F
さらに表示
FormPowder or granular powder
Assay from Suppliers CofAFluoride: 31.57-32.20%
Assay from Suppliers CofA≥99.0% (based on fluoride)
Appearance (Color)White to grey or pale brown
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of Freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only