Thermo Scientific Chemicals

Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals

Catalog number: 011537.30
250 g, Each
Thermo Scientific Chemicals

Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals

Catalog number: 011537.30
250 g, Each
Quantity
Catalog number: 011537.30
also known as 011537-30
Price (USD)
Price: 71.70
Online price: 60.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7488-55-3
IUPAC Name
λ²-tin(2+) sulfate
Molecular Formula
O4SSn
InChI Key
OBBXFSIWZVFYJR-UHFFFAOYSA-L
SMILES
[Sn++].[O-]S([O-])(=O)=O
Assay (unspecified)
95.5% min

Description

It is used in tin plating baths and it is a precursor for tin metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text